2,5-Dibromophenylacetic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(2,5-Dibromophenyl)acetic acid

OR41182-1G OR41182-25G OR41182-5G
APOSOR41182-1GEA 21.9 EUR
APOSOR41182-1G APOSOR41182-25G APOSOR41182-5G
2,5-Dibromophenylacetic acid
2,5-Dibromophenylacetic acid
Formel: C₈H₆Br₂O₂
Molecular Weight: 293,95 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02664691
CAS-Nummer: 203314-28-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR