4-Iodsalicylsäure 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Hydroxy-4-iodobenzoic acid

OR41184-25G OR41184-5G OR41184-100G
APOSOR41184-25GEA 159 EUR
APOSOR41184-25G APOSOR41184-5G APOSOR41184-100G
4-Iodsalicylsäure 98%
4-Iodsalicylsäure
Formel: C₇H₅IO₃
Molecular Weight: 264,02 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00061127
CAS-Nummer: 16870-28-3
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR