2-(Phenylsulfanyl)anilin
Lieferant: Apollo Scientific
|
Achtung
|
OR4141-100G
OR4141-500G
APOSOR4141-100GEA
0
EUR
APOSOR4141-100G
APOSOR4141-500G
2-(Phenylsulfanyl)anilin
2-Aminophenyl phenyl sulphide
Formel:
C₁₂H₁₁NS Schmelzpunkt: 43 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00190139 CAS-Nummer: 1134-94-7 UN: 3077 ADR: 9+EHS,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...