5-Hydroxymethyluracil

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-(Hydroxymethyl)pyrimidine-2,4(1H,3H)-dione

OR4151-25G OR4151-5G OR4151-100G
APOSOR4151-25GEA 102 EUR
APOSOR4151-25G APOSOR4151-5G APOSOR4151-100G
5-Hydroxymethyluracil
5-Hydroxymethyluracil
Formel: C₅H₆N₂O₃
Molecular Weight: 142,114 g/mol
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006070
CAS-Nummer: 4433-40-3
EINECS: 224-636-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR