1-(2-Hydroxyphenyl)piperazin 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: o-(1-Piperazinyl)phenol

OR4153-10G OR4153-250G OR4153-50G
APOSOR4153-10GEA 81.2 EUR
APOSOR4153-10G APOSOR4153-250G APOSOR4153-50G
1-(2-Hydroxyphenyl)piperazin 98%
1-(2-Hydroxyphenyl)piperazin
Formel: C₁₀H₁₄N₂O
Molecular Weight: 178,23 g/mol
Schmelzpunkt: 125…129 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00190246
CAS-Nummer: 1011-17-2
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR