9-Phenylcarbazole-3-boronic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: (9-Phenyl-9H-carbazol-3-yl)boronic acid, 9-Phenyl-3-carbazoleboronic acid

OR42024-25G OR42024-5G
APOSOR42024-25GEA 58 EUR
APOSOR42024-25G APOSOR42024-5G
9-Phenylcarbazole-3-boronic acid
9-Phenylcarbazole-3-boronic acid
Formel: C₁₈H₁₄BNO₂
Molecular Weight: 287.13 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD12196936
CAS-Nummer: 854952-58-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR