6-Brom-3-isochinolinol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Bromo-3-hydroxyisoquinoline
OR42117-1G
OR42117-500MG
APOSOR42117-1GEA
29
EUR
APOSOR42117-1G
APOSOR42117-500MG
6-Brom-3-isochinolinol
6-Brom-3-isochinolinol
Formel:
C₉H₆BrNO Molecular Weight: 224,0500 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD11110423 CAS-Nummer: 1031927-91-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...