2-Methylquinoline-4-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR4212-1G OR4212-5G
APOSOR4212-1GEA 21.9 EUR
APOSOR4212-1G APOSOR4212-5G
2-Methylquinoline-4-carboxylic acid
2-Methylquinoline-4-carboxylic acid
Formel: C₁₁H₉NO₂
Molecular Weight: 187.2 g/mol
Schmelzpunkt: 245…247 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00093919
CAS-Nummer: 634-38-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR