1,3-Benzoxazol-2-amin 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Benzo[d]oxazol-2-amine, 2-Aminobenzoxazole

OR42126-1G OR42126-10G OR42126-5G
APOSOR42126-1GEA 21.9 EUR
APOSOR42126-1G APOSOR42126-10G APOSOR42126-5G
1,3-Benzoxazol-2-amin 98%
1,3-Benzoxazol-2-amin
Formel: C₇H₆N₂O
Molecular Weight: 134.1353 g/mol
Schmelzpunkt: 129 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00182850
CAS-Nummer: 4570-41-6
EINECS: 224-952-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR