4-Brom-3-nitrophenol 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR42128-25G
OR42128-100G
APOSOR42128-25GEA
85.7
EUR
APOSOR42128-25G
APOSOR42128-100G
4-Brom-3-nitrophenol 98%
4-Brom-3-nitrophenol
Formel:
C₆H₄BrNO₃ Molecular Weight: 218,0100 g/mol Siedepunkt: 281 °C Schmelzpunkt: 144…145 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD11040194 CAS-Nummer: 78137-76-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...