5-Amino-2-chlor-3-methylpyridin

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Amino-2-chloro-3-picoline, 6-Chloro-5-methylpyridin-3-amine

OR42134-1G OR42134-5G
APOSOR42134-1GEA 21.9 EUR
APOSOR42134-1G APOSOR42134-5G
5-Amino-2-chlor-3-methylpyridin
5-Amino-2-chlor-3-methylpyridin
Formel: C₆H₇ClN₂
Molecular Weight: 142,5900 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03095087
CAS-Nummer: 38186-82-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR