Formamidin-hydrochlorid 96%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Formimidamide hydrochloride
OR42146-25G
OR42146-100G
OR42146-5G
APOSOR42146-25GEA
27.7
EUR
APOSOR42146-25G
APOSOR42146-100G
APOSOR42146-5G
Formamidin-hydrochlorid 96%
Formamidine hydrochloride
Formel:
CH₄N₂ Molecular Weight: 80,52 g/mol Schmelzpunkt: 84…87 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012865 CAS-Nummer: 6313-33-3 EINECS: 228-639-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...