3-Amino-4-methoxypyridin 95%

Lieferant: Apollo Scientific
Achtung

OR42156-1G OR42156-250MG
APOSOR42156-1GEA 24.8 EUR
APOSOR42156-1G APOSOR42156-250MG
3-Amino-4-methoxypyridin 95%
3-Amino-4-methoxypyridine
Formel: C₆H₈N₂O
Molecular Weight: 124,1400 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD07374959
CAS-Nummer: 33631-09-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR