2-Cyano-5-hydroxypyridine 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Hydroxypyridine-2-carbonitrile
OR42158-1G
OR42158-250MG
OR42158-5G
APOSOR42158-1GEA
102
EUR
APOSOR42158-1G
APOSOR42158-250MG
APOSOR42158-5G
2-Cyano-5-hydroxypyridine 97%
2-Cyano-5-hydroxypyridine
Formel:
C₆H₄N₂O Molecular Weight: 120,1100 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07368197 CAS-Nummer: 86869-14-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...