Methyl-2-amino-5-ethyl-1,3-thiazole-4-carboxylate

Lieferant: Apollo Scientific
Achtung

OR42181-1G OR42181-250MG OR42181-5G
APOSOR42181-1GEA 48 EUR
APOSOR42181-1G APOSOR42181-250MG APOSOR42181-5G
Methyl-2-amino-5-ethyl-1,3-thiazole-4-carboxylate
Methyl-2-amino-5-ethyl-1,3-thiazole-4-carboxylate
Formel: C₇H₁₀N₂O₂S
Molecular Weight: 186,23 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00728625
CAS-Nummer: 28942-54-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR