N-{[(2-Methyl-2-propanyl)oxy]carbonyl}-L-threonin

Lieferant: Apollo Scientific

Synonyme: N-Boc-L-threonine, Boc-Thr-OH, (2S,3R)-2-((tert-Butoxycarbonyl)amino)-3-hydroxybutanoic acid

OR42214-100G OR42214-25G OR42214-500G
APOSOR42214-100GEA 105 EUR
APOSOR42214-100G APOSOR42214-25G APOSOR42214-500G
N-{[(2-Methyl-2-propanyl)oxy]carbonyl}-L-threonin
N-Boc-L-threonine
Formel: C₉H₁₇NO₅
Molecular Weight: 219,2 g/mol
Schmelzpunkt: 80…82 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00065946
CAS-Nummer: 2592-18-9
EINECS: 219-987-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR