(S)-(+)-γ-Hydroxymethyl-γ-butyrolactone

Lieferant: Apollo Scientific
Achtung

Synonyme: (5S)-(+)-5-(Hydroxymethyl)dihydrofuran-2(3H)-one, (S)-5-(Hydroxymethyl)dihydrofuran-2(3H)-one, (S)-(+)-4,5-Dihydro-5-hydroxymethyl-2(3H)-furanone, (S)-(+)-5-Hydroxymethyltetrahydro-2-furanone

OR42217-25G OR42217-5G
APOSOR42217-25GEA 806 EUR
APOSOR42217-25G APOSOR42217-5G
(S)-(+)-γ-Hydroxymethyl-γ-butyrolactone
(S)-(+)-gamma-Hydroxymethyl-gamma-butyrolactone
Formel: C₅H₈O₃
Molecular Weight: 116,12 g/mol
Siedepunkt: 110…115 °C/0,2mmHg
Dichte: 1,237 g/cm³ (25 °C)
Flammpunkt: 113 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00066224
CAS-Nummer: 32780-06-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR