3-Chlor-4-(4-morpholinylcarbonyl)phenylboronsäure 98%
Lieferant: Apollo Scientific
Synonyme:
3-Chloro-4-(4-morpholinylcarbonyl)benzeneboronic acid, 3-Chloro-4-(morpholine-4-carbonyl)phenylboronic acid
OR4223-1G
OR4223-5G
APOSOR4223-1GEA
273
EUR
APOSOR4223-1G
APOSOR4223-5G
3-Chlor-4-(4-morpholinylcarbonyl)phenylboronsäure 98%
3-Chlor-4-(4-morpholinylcarbonyl)phenylboronsäure
Formel:
C₁₁H₁₃BClNO₄ Molecular Weight: 253,49 g/mol Schmelzpunkt: 182…186 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07363774 CAS-Nummer: 850589-49-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...