(9,9-Dimethyl-9H-xanthen-4,5-diyl)bis(diphenylphosphin)

Lieferant: Apollo Scientific
Achtung

Synonyme: XANTPHOS, 9,9-Dimethyl-4,5-bis(diphenylphosphino)xanthene, 4,5-Bis(diphenylphosphino)-9,9-dimethylxanthene

OR42230-25G OR42230-5G OR42230-100G
APOSOR42230-25GEA 63.8 EUR
APOSOR42230-25G APOSOR42230-5G APOSOR42230-100G
(9,9-Dimethyl-9H-xanthen-4,5-diyl)bis(diphenylphosphin)
(9,9-Dimethyl-9H-xanthene-4,5-diyl)bis(diphenylphosphine)
Formel: C₃₉H₃₂OP₂
Molecular Weight: 578,62 g/mol
Schmelzpunkt: 224…228 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00233866
CAS-Nummer: 161265-03-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR