Methyl-2-amino-4-brombenzoat
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Amino-4-bromobenzoic acid methyll ester, Methyl 2-amino-4-bromobenzoate
OR4230-100G
OR4230-25G
OR4230-5G
APOSOR4230-100GEA
194
EUR
APOSOR4230-100G
APOSOR4230-25G
APOSOR4230-5G
Methyl-2-amino-4-brombenzoat
Methyl-2-amino-4-brombenzoat
Formel:
C₈H₈BrNO₂ Molecular Weight: 230,0586 g/mol Schmelzpunkt: 77…79 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07783014 CAS-Nummer: 135484-83-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...