4-(Hydrazinocarbonyl)benzeneboronic acid 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(4-(Hydrazinecarbonyl)phenyl)boronic acid
OR4245-1G
OR4245-5G
APOSOR4245-1GEA
111
EUR
APOSOR4245-1G
APOSOR4245-5G
4-(Hydrazinocarbonyl)benzeneboronic acid 98%
4-(Hydrazinocarbonyl)benzeneboronic acid
Formel:
C₇H₉BN₂O₃ Molecular Weight: 179.97 g/mol Schmelzpunkt: 236…240 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD06659865 CAS-Nummer: 850567-95-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...