5-Chlor-2-hydroxyphenylboronsäure 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Chloro-2-hydroxybenzeneboronic acid

OR4250-1G OR4250-5G OR4250-250MG
APOSOR4250-1GEA 31.9 EUR
APOSOR4250-1G APOSOR4250-5G APOSOR4250-250MG
5-Chlor-2-hydroxyphenylboronsäure 97%
5-Chlor-2-hydroxyphenylboronsäure
Formel: C₆H₆BClO₃
Molecular Weight: 172,37 g/mol
Schmelzpunkt: 166…170 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06659863
CAS-Nummer: 89488-25-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR