2,6-Dichlorbenzoxazol 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR4251-25G
OR4251-5G
APOSOR4251-25GEA
24.8
EUR
APOSOR4251-25G
APOSOR4251-5G
2,6-Dichlorbenzoxazol 97%
2,6-Dichlorobenzoxazole
Formel:
C₇H₃Cl₂NO Molecular Weight: 188,0108 g/mol Siedepunkt: 110 °C Schmelzpunkt: 50 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07368635 CAS-Nummer: 3621-82-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...