3-Amino-1H-1,2,4-triazol-5-carbonsäure 96%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Amino-1,2,4-triazole-5-carboxylic acid Hemihydrate
OR4265-100G
OR4265-25G
OR4265-5G
APOSOR4265-100GEA
190
EUR
APOSOR4265-100G
APOSOR4265-25G
APOSOR4265-5G
3-Amino-1H-1,2,4-triazol-5-carbonsäure 96%
5-Amino-4H-1,2,4-triazole-3-carboxylic acid
Formel:
C₃H₄N₄O₂ Molecular Weight: 128.09 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010620 CAS-Nummer: 3641-13-2 EINECS: 222-868-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...