2-Amino-3,5-dichlorpyridin 97+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,5-Dichlorpyridin-2-amin, 3,5-Dichlor-2-pyridylamin

OR4268-25G OR4268-5G
APOSOR4268-25GEA 24.8 EUR
APOSOR4268-25G APOSOR4268-5G
2-Amino-3,5-dichlorpyridin 97+%
2-Amino-3,5-dichlorpyridin
Formel: C₅H₄Cl₂N₂
Molecular Weight: 163.01 g/mol
Schmelzpunkt: 81…83 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006313
CAS-Nummer: 4214-74-8
EINECS: 224-143-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR