5-Brom-2-hydroxypyridin

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Brom-6-pyridinol, 3-Brom-6-hydroxypyridin, 5-Brom-2-pyridinol

OR4273-25G OR4273-5G
APOSOR4273-25GEA 24.8 EUR
APOSOR4273-25G APOSOR4273-5G
5-Brom-2-hydroxypyridin
5-Brom-2-hydroxypyridin
Formel: C₅H₄BrNO
Molecular Weight: 174 g/mol
Schmelzpunkt: 174…178 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00234042
CAS-Nummer: 13466-38-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR