3-(2-Furyl)-1H-pyrazole

Lieferant: Apollo Scientific
Achtung

OR43-1G OR43-5G
APOSOR43-1GEA 79.9 EUR
APOSOR43-1G APOSOR43-5G
3-(2-Furyl)-1H-pyrazole
3-(2-Furyl)-1H-pyrazole
Formel: C₇H₆N₂O
Molecular Weight: 134.14 g/mol
Schmelzpunkt: 102…104 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00973923
CAS-Nummer: 32332-98-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR