4-Iodbenzonitril

Lieferant: Apollo Scientific
Achtung

Synonyme: p-Iodobenzonitrile

OR4305-100G OR4305-25G OR4305-500G
APOSOR4305-100GEA 91.5 EUR
APOSOR4305-100G APOSOR4305-25G APOSOR4305-500G
4-Iodbenzonitril
4-Iodbenzonitril
Formel: C₇H₄IN
Molecular Weight: 229,02 g/mol
Schmelzpunkt: 124…128 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00051310
CAS-Nummer: 3058-39-7
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR