1-Phenylvinylboronic acid pinacol ester 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4,4,5,5-Tetramethyl-2-(1-phenylvinyl)-1,3,2-dioxaborolane

OR4310-1G OR4310-5G
APOSOR4310-1GEA 49.3 EUR
APOSOR4310-1G APOSOR4310-5G
1-Phenylvinylboronic acid pinacol ester 96%
1-Phenylvinylboronic acid pinacol ester
Formel: C₁₄H₁₉BO₂
Molecular Weight: 230,11 g/mol
Schmelzpunkt: 42…49 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06659923
CAS-Nummer: 143825-84-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR