3-Brom-2-chlor-4-pyridinboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Brom-2-chlorpyridin-4-boronsäure, 3-Brom-2-chlor-4-pyridinylboronsäure

OR4323-1G OR4323-5G OR4323-25G
APOSOR4323-1GEA 60.9 EUR
APOSOR4323-1G APOSOR4323-5G APOSOR4323-25G
3-Brom-2-chlor-4-pyridinboronsäure
3-Bromo-2-chloro-4-pyridineboronic acid
Formel: C₅H₄BBrClNO₂
Molecular Weight: 236.26 g/mol
Schmelzpunkt: 171 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08458470
CAS-Nummer: 1072944-16-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR