4-Brom-1-chlor-2-ethylbenzol

Lieferant: Apollo Scientific
Achtung

OR4333-1G OR4333-25G OR4333-5G
APOSOR4333-1GEA 21.9 EUR
APOSOR4333-1G APOSOR4333-25G APOSOR4333-5G
4-Brom-1-chlor-2-ethylbenzol
4-Brom-1-chlor-2-ethylbenzol
Formel: C₈H₈BrCl
Molecular Weight: 219,51 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00672954
CAS-Nummer: 289039-22-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR