(7-Bromo-1-benzothiophen-2-yl)methanol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(7-Bromobenzo[b]thiophen-2-yl)methanol, 7-Bromo-2-(hydroxymethyl)benzo[b]thiophene
OR43532-100MG
OR43532-1G
OR43532-250MG
APOSOR43532-100MGEA
30.6
EUR
APOSOR43532-100MG
APOSOR43532-1G
APOSOR43532-250MG
(7-Bromo-1-benzothiophen-2-yl)methanol
(7-Bromo-1-benzothiophen-2-yl)methanol
Formel:
C₉H₇BrOS Molecular Weight: 243,13 g/mol Schmelzpunkt: 77…78 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD12032282 CAS-Nummer: 1171926-64-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...