6-Brom-4-chlorchinolin 98%

Lieferant: Apollo Scientific
Achtung

OR43545-1G OR43545-5G OR43545-10G
APOSOR43545-1GEA 21.9 EUR
APOSOR43545-1G APOSOR43545-5G APOSOR43545-10G
6-Brom-4-chlorchinolin 98%
6-Bromo-4-chloroquinoline
Formel: C₉H₅BrClN
Molecular Weight: 242.50 g/mol
Schmelzpunkt: 123…125 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00511001
CAS-Nummer: 65340-70-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR