4-Amino-3-brom-1-methyl-1H-indazol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Bromo-1-methyl-1H-indazol-4-amine
OR43549-1G
OR43549-250MG
APOSOR43549-1GEA
252
EUR
APOSOR43549-1G
APOSOR43549-250MG
4-Amino-3-brom-1-methyl-1H-indazol
4-Amino-3-bromo-1-methyl-1H-indazole
Formel:
C₈H₈BrN₃ Molecular Weight: 226.0700 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD11109355 CAS-Nummer: 1092351-47-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...