5-Amino-3-brom-1-methyl-1H-indazol

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Bromo-1-methyl-1H-indazol-5-amine

OR43550-1G OR43550-250MG
APOSOR43550-1GEA 27.7 EUR
APOSOR43550-1G APOSOR43550-250MG
5-Amino-3-brom-1-methyl-1H-indazol
5-Amino-3-bromo-1-methyl-1H-indazole
Formel: C₈H₈BrN₃
Molecular Weight: 226,0700 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11100026
CAS-Nummer: 1092351-49-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR