5-Brom-1,7-dimethyl-1H-indazol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Bromo-1,7-dimethylindazole
OR43557-1G
OR43557-250MG
APOSOR43557-1GEA
92.8
EUR
APOSOR43557-1G
APOSOR43557-250MG
5-Brom-1,7-dimethyl-1H-indazol
5-Bromo-1,7-dimethyl-1H-indazole
Formel:
C₉H₉BrN₂ Molecular Weight: 225.09 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD11226595 CAS-Nummer: 1092352-34-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...