5-Brom-1H-pyrazolo[3,4-b]pyridin

Lieferant: Apollo Scientific
Achtung

OR43562-500MG OR43562-5G OR43562-25G
APOSOR43562-500MGEA 21.9 EUR
APOSOR43562-500MG APOSOR43562-5G APOSOR43562-25G
5-Brom-1H-pyrazolo[3,4-b]pyridin
5-Bromo-1H-pyrazolo[3,4-b]pyridine
Formel: C₆H₄BrN₃
Molecular Weight: 198,02 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05663982
CAS-Nummer: 875781-17-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR