6-Brom-1-methyl-1H-indazol 98%

Lieferant: Apollo Scientific
Achtung

OR43569-1G OR43569-5G OR43569-10G
APOSOR43569-1GEA 23.2 EUR
APOSOR43569-1G APOSOR43569-5G APOSOR43569-10G
6-Brom-1-methyl-1H-indazol 98%
6-Bromo-1-methyl-1H-indazole
Formel: C₈H₇BrN₂
Molecular Weight: 211.0600 g/mol
Schmelzpunkt: 48…50 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09878569
CAS-Nummer: 590417-94-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR