Methylimidazo[1,2-a]pyridin-5-carboxylat 98%

Lieferant: Apollo Scientific
Achtung

OR43572-1G OR43572-5G
APOSOR43572-1GEA 85.7 EUR
APOSOR43572-1G APOSOR43572-5G
Methylimidazo[1,2-a]pyridin-5-carboxylat 98%
Methylimidazo[1,2-a]pyridin-5-carboxylat
Formel: C₉H₈N₂O₂
Molecular Weight: 176.1700 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD12026020
CAS-Nummer: 88047-55-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR