1-Methyl-1H-pyrazole-4-boronic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: (1-Methyl-1H-pyrazol-4-yl)boronic acid

OR43581-1G OR43581-5G OR43581-10G
APOSOR43581-1GEA 21.9 EUR
APOSOR43581-1G APOSOR43581-5G APOSOR43581-10G
1-Methyl-1H-pyrazole-4-boronic acid
1-Methyl-1H-pyrazole-4-boronic acid
Formel: C₄H₇BN₂O₂
Molecular Weight: 125,92 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09414709
CAS-Nummer: 847818-55-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR