2,4,5-Trichlorbenzoesäure

Lieferant: Apollo Scientific
Achtung

OR43584-1G OR43584-25G OR43584-5G
APOSOR43584-1GEA 21.9 EUR
APOSOR43584-1G APOSOR43584-25G APOSOR43584-5G
2,4,5-Trichlorbenzoesäure
2,4,5-Trichlorobenzoic acid
Formel: C₇H₃Cl₃O₂
Molecular Weight: 225.4600 g/mol
Schmelzpunkt: 159…164 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01662044
CAS-Nummer: 50-82-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR