5-Bromo-6-chloro-2-methyl-4-pyrimidinol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Bromo-6-chloro-4-hydroxy-2-methylpyrimidine
OR43598-1G
OR43598-500MG
OR43598-100MG
APOSOR43598-1GEA
255
EUR
APOSOR43598-1G
APOSOR43598-500MG
APOSOR43598-100MG
5-Bromo-6-chloro-2-methyl-4-pyrimidinol
5-Bromo-6-chloro-2-methyl-4-pyrimidinol
Formel:
C₅H₄BrClN₂O Molecular Weight: 223,46 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11840977 CAS-Nummer: 105806-11-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...