4-Amino-6-methyl-2H-pyridazin-3-one 97%

Lieferant: Apollo Scientific
Achtung

OR43619-10G OR43619-1G OR43619-5G
APOSOR43619-10GEA 1410 EUR
APOSOR43619-10G APOSOR43619-1G APOSOR43619-5G
4-Amino-6-methyl-2H-pyridazin-3-one 97%
4-Amino-6-methyl-2H-pyridazin-3-one
Formel: C₅H₇N₃O
Molecular Weight: 125,13 g/mol
Schmelzpunkt: 257…261 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00183934
CAS-Nummer: 13925-21-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR