4-Brom-2,6-dichlorpyridin 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR43622-1G
OR43622-5G
OR43622-10G
APOSOR43622-1GEA
50.9
EUR
APOSOR43622-1G
APOSOR43622-5G
APOSOR43622-10G
4-Brom-2,6-dichlorpyridin 97%
4-Bromo-2,6-dichloropyridine
Formel:
C₅H₂BrCl₂N Molecular Weight: 226,8900 g/mol Schmelzpunkt: 95…102 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11848646 CAS-Nummer: 98027-80-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...