4-Amino-1-methyl-1H-pyrazol Hydrochlorid 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Methyl-1H-pyrazol-4-amine hydrochloride
OR43645-25G
OR43645-100G
APOSOR43645-25GEA
58
EUR
APOSOR43645-25G
APOSOR43645-100G
4-Amino-1-methyl-1H-pyrazol Hydrochlorid 95%
4-Amino-1-methyl-1H-pyrazol Hydrochlorid
Formel:
C₄H₈ClN₃ Molecular Weight: 133,5794 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07626616 CAS-Nummer: 127107-23-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...