3-(Bromomethyl)-2-chlorothiophene 97%

Lieferant: Apollo Scientific
Gefahr

OR43664-1G OR43664-5G
APOSOR43664-1GEA 116 EUR
APOSOR43664-1G APOSOR43664-5G
3-(Bromomethyl)-2-chlorothiophene 97%
3-(Bromomethyl)-2-chlorothiophene
Formel: C5H4BrClS
Molecular Weight: 211,51 g/mol
Siedepunkt: 88 °C/1mmHg
Lagertemperatur: Kühlschrank
MDL: MFCD06411087
CAS-Nummer: 40032-81-3
UN: 3265
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR