(±)-4-Brommandelsäure 96+%

Lieferant: Apollo Scientific
Achtung

Synonyme: (±)-2-(4-Bromphenyl)-2-hydroxyessigsäure, (±)-4-Bromphenylglykolsäure, (±)-2-(4-Bromphenyl)-2-hydroxyethansäure

OR43666-100G OR43666-25G OR43666-5G
APOSOR43666-100GEA 134 EUR
APOSOR43666-100G APOSOR43666-25G APOSOR43666-5G
(±)-4-Brommandelsäure 96+%
(±)-4-Brommandelsäure
Formel: C₈H₇BrO₃
Molecular Weight: 231,05 g/mol
Schmelzpunkt: 115…119 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004232
CAS-Nummer: 6940-50-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR