4-(Benzyloxy)phenol 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Monobenzon, Hydrochinonmonobenzylether, 4-Hydroxyphenylbenzylether
OR4376-500G
APOSOR4376-500GEA
194
EUR
APOSOR4376-500G
4-(Benzyloxy)phenol 98%
4-Benzyloxyphenol
Formel:
C₁₃H₁₂O₂ Molecular Weight: 200,24 g/mol Schmelzpunkt: 118…122 °C Dichte: 1,26 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002333 CAS-Nummer: 103-16-2 EINECS: 203-083-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...