1-Acetyl-5-indolinesulphonyl chloride 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 1-acetyl-2,3-dihydro-1H-indole-5-sulphonyl chloride, n-acetylindoline-5-sulphonyl chloride, 1-acetylindoline-5-sulphonyl chloride

OR4396-1G OR4396-250MG OR4396-5G
APOSOR4396-1GEA 59.6 EUR
APOSOR4396-1G APOSOR4396-250MG APOSOR4396-5G
1-Acetyl-5-indolinesulphonyl chloride 97%
1-Acetyl-5-indolinesulphonyl chloride
Formel: C₁₀H₁₀ClNO₃S
Molecular Weight: 259,71 g/mol
Schmelzpunkt: 172…174 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD07368558
CAS-Nummer: 52206-05-0
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR