6-Nitrochinoxalin

Lieferant: Apollo Scientific
Gefahr

OR4401-1G OR4401-5G OR4401-25G
APOSOR4401-1GEA 21.9 EUR
APOSOR4401-1G APOSOR4401-5G APOSOR4401-25G
6-Nitrochinoxalin
6-Nitroquinoxaline
Formel: C₈H₅N₃O₂
Molecular Weight: 175,15 g/mol
Schmelzpunkt: 175…179 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00462822
CAS-Nummer: 6639-87-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR